Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkacb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164198.1 | NP_001157670.1 | 10 | 2435 | ||
NM_001164199.1 | NP_001157671.1 | 10 | 2308 | |||
NM_001164200.1 | NP_001157672.1 | 10 | 2400 | |||
NM_011100.4 | NP_035230.1 | 10 | 2439 | |||
GenBank mRNA | AK035560.1 | - | 3 | 1523 | ||
AK048319.1 | - | 10 | 2431 | |||
AK081984.1 | - | 3 | 1449 | |||
AK139419.1 | - | 10 | 2180 | |||
AK148728.1 | - | 10 | 2249 | |||
AK150122.1 | - | 10 | 2401 | |||
AK150134.1 | - | 10 | 2401 | |||
AK163585.1 | - | 10 | 2214 | |||
BC054533.1 | - | 10 | 2423 | 5704796 |