Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011149.2 | NP_035279.2 | 1 | 223 | ||
GenBank mRNA | AK002357.1 | - | 1 | 170 | ||
AK128977.1 | - | 1 | 173 | |||
AK154073.1 | - | 1 | 162 | |||
AL363449.1 | - | 1 | 264 | |||
BC013061.1 | - | 1 | 160 | 3483267 | ||
M60456.1 | - | 1 | 142 | |||
X58990.1 | - | 1 | 268 |