Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyth1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112699.1 | NP_001106169.1 | 13 | 1738 | ||
NM_001112700.1 | NP_001106170.1 | 14 | 1681 | |||
NM_011180.3 | NP_035310.2 | 14 | 1741 | |||
GenBank mRNA | AB035538.1 | - | 13 | 1710 | ||
AK158216.1 | - | 14 | 1681 | |||
AK172296.1 | - | 14 | 1686 | |||
AK220302.1 | - | 14 | 1663 | |||
AY126516.1 | - | 13 | 1707 | |||
BC006658.1 | - | 1 | 1412 | |||
BC057974.1 | - | 13 | 1622 | 5060913 |