Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pth1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083935.1 | NP_001077404.1 | 3, 4 | 460 | ||
NM_001083936.1 | NP_001077405.1 | 2, 3 | 493 | |||
NM_011199.2 | NP_035329.2 | 4, 5 | 462 | |||
GenBank mRNA | BB614354.1 | - | 2, 3 | 494 | ||
BC013446.1 | - | 4, 5 | 439 | 4241234 | ||
BC051981.1 | - | 2, 3 | 364 | 6402767 | ||
CB192926.1 | - | 3, 4 | 422 | |||
X78936.1 | - | 2, 3 | 239 |
Set Membership: |
G-protein coupled receptor |