Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011213.2 | NP_035343.2 | 5 | 648 | ||
GenBank mRNA | AF300943.1 | - | 3 | 311 | ||
AI385919.1 | - | 1 | 29 | |||
BU056582.1 | - | 5 | 530 |