Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282848.1 | NP_001269777.1 | 8 | 1989 | ||
NM_001282849.1 | NP_001269778.1 | 7 | 1822 | |||
NM_004683.5 | NP_004674.1 | 7 | 1418 | |||
NM_152869.3 | NP_690608.1 | 8 | 2038 | |||
XM_006724567.2 | XP_006724630.1 | 6 | 1138 | |||
XM_006724568.2 | XP_006724631.1 | 5 | 1006 | |||
XM_017029954.1 | XP_016885443.1 | 6 | 1746 | |||
GenBank mRNA | AB028125.1 | - | 7 | 1142 | ||
AB032064.1 | - | 7 | 1200 | |||
AK223360.1 | - | 7 | 1418 | |||
AK290136.1 | - | 7 | 1356 | |||
BC050371.1 | - | 7 | 1760 | 5760604 | ||
BC073173.1 | - | 7 | 1232 | 6186669 | ||
D31815.1 | - | 7 | 1141 | |||
EU794655.1 | - | 7 | 1200 |