Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011247.2 | NP_035377.2 | 11 | 1757 | ||
GenBank mRNA | U28789.1 | - | 5 | 751 | ||
U83913.1 | - | 6 | 900 |