Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011421.2 | NP_035551.1 | 2 | 1215 | ||
GenBank mRNA | AK088147.1 | - | 2 | 1184 | ||
AK138133.1 | - | 2 | 1149 | |||
AK145534.1 | - | 2 | 1184 | |||
AK145702.1 | - | 2 | 1216 | |||
AK149579.1 | - | 2 | 1184 | |||
AK164167.1 | - | 2 | 1184 | |||
AK164717.1 | - | 2 | 1139 | |||
AK167802.1 | - | 2 | 1138 | |||
AK190981.1 | - | 1 | 249 | |||
BC011304.1 | - | 2 | 1150 | 3964394 | ||
BC034515.1 | - | 2 | 1140 | 4482098 | ||
Z14252.1 | - | 2 | 1161 |