Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCPG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204450.1 | NP_001191379.1 | 3, 4 | 479 | ||
NM_001204451.1 | NP_001191380.1 | 3, 4 | 479 | |||
NM_004748.4 | NP_004739.3 | 3, 4 | 719 | |||
NM_020739.3 | NP_065790.2 | 3, 4 | 479 | |||
GenBank mRNA | AB033080.2 | - | 3, 4 | 264 | ||
AF212228.1 | - | 3, 4 | 395 | |||
AK022459.1 | - | 3, 4 | 275 | |||
AK292795.1 | - | 3, 4 | 228 | |||
AL705964.1 | - | 3, 4 | 424 | |||
BC015203.1 | - | 3, 4 | 315 | |||
BC039871.1 | - | 3, 4 | 233 | 4385087 | ||
DB084778.1 | - | 3, 4 | 479 |
Gene Symbol: DYX1C1-CCPG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037923.1 | 10, 11 | 1597 | |||
GenBank mRNA | AK295788.1 | - | 8, 9 | 1094 |