Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sox6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277326.1 | NP_001264255.1 | 9 | 1137 | ||
NM_011445.4 | NP_035575.1 | 10 | 1190 | |||
GenBank mRNA | AK132409.1 | - | 9 | 1103 | ||
U32614.1 | - | 10 | 1194 |