Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP2K3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316332.1 | NP_001303261.1 | 3 | 460 | ||
NM_002756.4 | NP_002747.2 | 2 | 77 | |||
NM_145109.2 | NP_659731.1 | 2 | 335 | |||
XM_005256723.2 | XP_005256780.1 | 3 | 375 | |||
XM_011523958.2 | XP_011522260.1 | 2 | 134 | |||
XM_011523959.1 | XP_011522261.1 | 4 | 342 | |||
XM_017024857.1 | XP_016880346.1 | 2 | 204 | |||
XM_017024858.1 | XP_016880347.1 | 3 | 192 | |||
XM_017024859.1 | XP_016880348.1 | 4 | 532 | |||
GenBank mRNA | AB451226.1 | - | 2 | 70 | ||
AB451349.1 | - | 2 | 70 | |||
AK093838.1 | - | 3 | 379 | |||
AK223490.1 | - | 2 | 210 | |||
AK315505.1 | - | 2 | 280 | |||
BC032478.1 | - | 2 | 270 | 5215093 | ||
BM836867.1 | - | 2 | 315 | |||
CR536514.1 | - | 2 | 70 | |||
CR541793.1 | - | 2 | 70 | |||
D87116.1 | - | 2 | 127 | |||
DA131175.1 | - | 2 | 335 | |||
DA987047.1 | - | 2 | 78 | |||
GQ225578.1 | - | 2 | 70 | |||
L36719.1 | - | 4 | 320 | |||
U66839.1 | - | 2 | 196 |