Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stxbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011504.1 | NP_035634.1 | 7 | 504 | ||
GenBank mRNA | AK138609.1 | - | 7 | 536 | ||
AK151466.1 | - | 7 | 499 | |||
AK152261.1 | - | 7 | 493 | |||
AK155133.1 | - | 7 | 502 | |||
AK169799.1 | - | 7 | 505 | |||
BC062901.1 | - | 7 | 513 | 6810254 | ||
BC094055.1 | - | 7 | 512 | |||
D30798.1 | - | 7 | 504 | |||
U19521.1 | - | 7 | 629 |