Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NREP SIRNA Assays ›
Gene Symbol:
NREP
Gene Name:
neuronal regeneration related protein
Gene Aliases:
C5orf13, D4S114, P311, PRO1873, PTZ17, SEZ17
Chromosome Location:
Chr.5: 111729303 - 111976931 on Build GRCh38
UniGene ID:
Hs.36053
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
15191
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NREP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142474.1
NP_001135946.1
3, 4
373
NM_001142475.1
NP_001135947.1
3, 4
403
NM_001142476.1
NP_001135948.1
4, 5
455
NM_001142477.1
NP_001135949.1
4, 5
346
NM_001142478.1
NP_001135950.1
4, 5
322
NM_001142479.1
NP_001135951.1
4, 5
359
NM_001142480.1
NP_001135952.1
4, 5
353
NM_001142481.1
NP_001135953.1
4, 5
262
NM_001142482.1
NP_001135954.1
4, 5
269
NM_001142483.1
NP_001135955.1
2, 3
558
NM_004772.2
NP_004763.1
3, 4
250
XM_006714732.1
XP_006714795.1
2, 3
170
XM_006714733.2
XP_006714796.1
3, 4
151
GenBank mRNA
AK298779.1
-
3, 4
298
AK315617.1
-
3, 4
224
AL547663.3
-
4, 5
350
BC011050.1
-
4, 5
283
4340005
BC019068.2
-
3, 4
186
4893075
BC072013.1
-
3, 4
209
5922876
BC072443.1
-
3, 4
214
6162335
DA027062.1
-
3, 4
373
DA078027.1
-
2, 3
283
DA490948.1
-
4, 5
455
DA520221.1
-
4, 5
281
DA526263.1
-
4, 5
443
DA534424.1
-
2, 3
558
DA683910.1
-
4, 5
269
DA694580.1
-
3, 4
268
DA704824.1
-
4, 5
359
DA744980.1
-
4, 5
206
DA752769.1
-
4, 5
262
DA908653.1
-
3, 4
250
DA939543.1
-
4, 5
346
DC365764.1
-
3, 4
403
U30521.1
-
4, 5
281
U36189.1
-
2, 3
94
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transforming growth factor beta receptor signaling pathway
axon regeneration
regulation of neuron differentiation
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0