Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3gl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252471.1 | NP_001239400.1 | 4 | 382 | ||
NM_013664.3 | NP_038692.1 | 4 | 382 | |||
GenBank mRNA | AK154836.1 | - | 4 | 381 | ||
AK162715.1 | - | 4 | 310 | |||
AK167505.1 | - | 4 | 324 | |||
BC138602.1 | - | 4 | 321 | 8861622 | ||
BC138603.1 | - | 4 | 321 | 8861623 | ||
BC145345.1 | - | 4 | 321 | |||
U58885.1 | - | 4 | 321 |