Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004791.2 | NP_004782.1 | 2, 3 | 565 | ||
XM_005254100.4 | XP_005254157.1 | 2, 3 | 523 | |||
GenBank mRNA | AB008375.1 | - | 3, 4 | 428 | ||
AF072752.1 | - | 2, 3 | 543 | |||
AK095875.1 | - | 2, 3 | 205 | |||
AK291340.1 | - | 2, 3 | 485 | |||
BC036788.1 | - | 2, 3 | 486 | 5753239 |