Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016795.3 | NP_058075.2 | 5 | 411 | ||
GenBank mRNA | AB012290.1 | - | 4 | 369 | ||
AJ224115.1 | - | 5 | 405 | |||
AK015754.1 | - | 2 | 153 | |||
AK017187.1 | - | 5 | 408 | |||
AK141271.1 | - | 5 | 478 | |||
AK151152.1 | - | 5 | 474 | |||
AK161346.1 | - | 5 | 389 | |||
AK161388.1 | - | 5 | 390 | |||
BC005707.1 | - | 5 | 380 | 3157835 | ||
BC050761.1 | - | 5 | 411 | 6771151 |