Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017462.2 | NP_059490.2 | 9 | 1929 | ||
GenBank mRNA | AB121698.1 | - | 8 | 1622 | ||
AK133584.1 | - | 9 | 1855 | |||
AK148091.1 | - | 9 | 1891 | |||
BC053024.1 | - | 9 | 1855 | |||
BC061472.1 | - | 9 | 1843 | 6827238 | ||
BC080291.1 | - | 9 | 1760 | |||
U53584.1 | - | 9 | 1849 |