Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019647.6 | NP_062621.2 | 6 | 1274 | ||
GenBank mRNA | AK018480.1 | - | 6 | 1020 | ||
BC138299.1 | - | 6 | 1025 | 8861318 | ||
BC138300.1 | - | 6 | 1025 | 8861319 |