Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019755.4 | NP_062729.1 | 5 | 755 | ||
GenBank mRNA | AK003522.1 | - | 5 | 810 | ||
AK011282.1 | - | 5 | 809 | |||
AK012816.1 | - | 4 | 942 | |||
AK154228.1 | - | 5 | 819 | |||
BC083078.1 | - | 5 | 798 | 6477560 | ||
BC092095.1 | - | 5 | 774 | 3989985 | ||
BC119088.1 | - | 5 | 755 | 8733837 | ||
BC119090.1 | - | 5 | 755 | 8733839 |