Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020600.4 | NP_065625.2 | 2 | 220 | ||
GenBank mRNA | AK003148.1 | - | 2 | 165 | ||
AK011213.1 | - | 2 | 165 | |||
AK012425.1 | - | 2 | 164 | |||
AK012584.1 | - | 2 | 164 | |||
AK019233.1 | - | 2 | 163 | |||
AK028106.1 | - | 2 | 168 | |||
AK146325.1 | - | 2 | 161 | |||
AK146976.1 | - | 2 | 164 | |||
AK152461.1 | - | 1 | 1883 | |||
AK153401.1 | - | 1 | 1883 | |||
AK168237.1 | - | 2 | 164 | |||
BC042940.1 | - | 1 | 144 | 6487681 | ||
BC062874.1 | - | 1 | 143 | 5683900 | ||
BC081449.1 | - | 1 | 131 | 5684375 | ||
BY149058.1 | - | 2 | 221 |