Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYMPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004819.2 | NP_004810.2 | 7 | 874 | ||
XM_005259286.1 | XP_005259343.1 | 7 | 827 | |||
XM_011527354.1 | XP_011525656.1 | 8 | 877 | |||
GenBank mRNA | AB209024.1 | - | 7 | 3122 | ||
AB210010.1 | - | 7 | 719 | |||
AK300252.1 | - | 7 | 728 | |||
BC006536.2 | - | 6 | 581 | |||
BC006567.2 | - | 6 | 581 | |||
BC030214.1 | - | 7 | 716 | |||
BX649165.1 | - | 7 | 729 | |||
U49240.1 | - | 2 | 343 | |||
Y10931.1 | - | 7 | 692 |