Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ryr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023868.2 | NP_076357.2 | 6 | 858 | ||
GenBank mRNA | AF295105.1 | - | 6 | 359 | ||
AK020368.1 | - | 5 | 580 | |||
AK086037.1 | - | 1 | 1711 | |||
BC043140.1 | - | 5 | 837 | |||
BC059061.1 | - | 5 | 837 |