Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133670.1 | NP_598431.1 | 4, 5 | 428 | ||
GenBank mRNA | AK002700.1 | - | 4, 5 | 451 | ||
BC005413.1 | - | 4, 5 | 428 | 3488459 | ||
L02331.1 | - | 4, 5 | 444 |