Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL27RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004843.3 | NP_004834.1 | 6 | 883 | ||
GenBank mRNA | AF053004.1 | - | 6 | 888 | ||
AF106912.1 | - | 6 | 875 | |||
AF265242.1 | - | 6 | 861 | |||
AY358876.1 | - | 6 | 861 | |||
BC028003.1 | - | 6 | 806 | 5212139 |