Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hps6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176785.3 | NP_789742.2 | 1 | 1643 | ||
GenBank mRNA | AF536239.1 | - | 1 | 1588 | ||
AK030582.1 | - | 1 | 1614 | |||
AK040849.1 | - | 1 | 1590 | |||
AK159588.1 | - | 1 | 1609 | |||
AK172058.1 | - | 2 | 1394 | |||
BC034055.1 | - | 1 | 1589 | 3673655 |