Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbbp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009031.3 | NP_033057.3 | 12 | 1813 | ||
GenBank mRNA | AK076016.1 | - | 12 | 1738 | ||
AK078349.1 | - | 12 | 1726 | |||
AK082892.1 | - | 12 | 1725 | |||
AK135779.1 | - | 12 | 1727 | |||
AK135869.1 | - | 11 | 1422 | |||
AK135875.1 | - | 11 | 1422 | |||
AK135956.1 | - | 12 | 1727 | |||
AK136110.1 | - | 12 | 1725 | |||
AK145531.1 | - | 12 | 1724 | |||
AK145651.1 | - | 12 | 1707 | |||
AK146014.1 | - | 12 | 1725 | |||
AK146439.1 | - | 12 | 1784 | |||
AK146904.1 | - | 12 | 1726 | |||
AK146967.1 | - | 12 | 1725 | |||
AK147062.1 | - | 12 | 1784 | |||
AK148852.1 | - | 12 | 1753 | |||
AK153913.1 | - | 12 | 1770 | |||
AK160023.1 | - | 12 | 1725 | |||
AK181761.1 | - | 1 | 264 | |||
AK206140.1 | - | 1 | 221 | |||
AK208251.1 | - | 1 | 200 | |||
AK213569.1 | - | 1 | 206 | |||
BC003785.1 | - | 12 | 1742 | 3600013 |