Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tox4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023434.3 | NP_075923.2 | 5 | 774 | ||
GenBank mRNA | AF228408.1 | - | 3 | 433 | ||
AK041829.1 | - | 3 | 1337 | |||
AK042459.1 | - | 3 | 1337 | |||
AK088144.1 | - | 4 | 446 | |||
AK147023.1 | - | 4 | 480 | |||
AK147834.1 | - | 4 | 451 | |||
AK153278.1 | - | 4 | 440 | |||
AK173012.1 | - | 4 | 478 | |||
AK195528.1 | - | 2 | 137 | |||
AK203209.1 | - | 2 | 137 | |||
AK207200.1 | - | 2 | 137 | |||
BC050091.1 | - | 3 | 429 | 30023547 |