Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec22a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133704.4 | NP_598465.2 | 7 | 3086 | ||
GenBank mRNA | AK082516.1 | - | 7 | 3087 | ||
AK085223.1 | - | 1 | 1660 | |||
AK161238.1 | - | 1 | 615 | |||
AK164647.1 | - | 1 | 1659 |