Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162492.1 | NP_001155964.1 | 3 | 815 | ||
NM_001162493.1 | NP_001155965.1 | 3 | 785 | |||
NM_138670.3 | NP_619611.3 | 3 | 919 | |||
GenBank mRNA | AK136571.1 | - | 2 | 814 | ||
AY974575.1 | - | 1 | 81 | |||
BC004079.1 | - | 2 | 760 | 3592659 | ||
BC006066.1 | - | 2 | 760 | |||
BC094469.1 | - | 3 | 919 | 5009481 |