Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139227.4 | NP_631973.3 | 8 | 1365 | ||
GenBank mRNA | AF455111.1 | - | 7 | 1105 | ||
AK046679.1 | - | 8 | 1366 | |||
BC035459.1 | - | 7 | 1018 | |||
BC137846.1 | - | 8 | 1286 | 8860865 |