Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC24A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301031.1 | NP_001287960.1 | 1 | 682 | ||
NM_001301032.1 | NP_001287961.1 | 1 | 682 | |||
NM_001301033.1 | NP_001287962.1 | 4 | 1180 | |||
NM_004727.2 | NP_004718.1 | 2 | 843 | |||
XM_005254778.3 | XP_005254835.1 | 3 | 817 | |||
XM_005254781.3 | XP_005254838.1 | 3 | 817 | |||
XM_006720768.3 | XP_006720831.1 | 3 | 817 | |||
XM_011522219.1 | XP_011520521.1 | 3 | 911 | |||
XM_011522220.2 | XP_011520522.1 | 3 | 817 | |||
XM_011522221.2 | XP_011520523.1 | 3 | 817 | |||
XM_011522222.2 | XP_011520524.1 | 3 | 817 | |||
XM_011522226.2 | XP_011520528.1 | 3 | 817 | |||
XM_017022724.1 | XP_016878213.1 | 3 | 817 | |||
XM_017022725.1 | XP_016878214.1 | 3 | 817 | |||
GenBank mRNA | AB014602.1 | - | 3 | 2031 | ||
AF026132.1 | - | 1 | 556 | |||
AF062921.1 | - | 1 | 556 | |||
AF062922.1 | - | 2 | 843 | |||
AK304006.1 | - | 4 | 1180 | |||
BC117263.1 | - | 1 | 638 | 40125814 | ||
BC143375.1 | - | 1 | 638 | 9051880 |