Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145146.2 | NP_660128.2 | 4 | 372 | ||
GenBank mRNA | AJ011080.1 | - | 4 | 400 | ||
AK143987.1 | - | 4 | 373 | |||
BC026681.1 | - | 4 | 290 | |||
BC100597.1 | - | 4 | 372 | 6767654 |