Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145589.2 | NP_663564.2 | 9, 10 | 1832 | ||
GenBank mRNA | AK133884.1 | - | 9, 10 | 1796 | ||
AK139559.1 | - | 10, 11 | 1607 | |||
AK145816.1 | - | 10, 11 | 1612 | |||
AK151203.1 | - | 10, 11 | 1664 | |||
AK182875.1 | - | 3, 4 | 271 | |||
AK207496.1 | - | 2, 3 | 189 | |||
BC055033.1 | - | 9, 10 | 1832 | 6310272 |