Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGFR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000142.4 | NP_000133.1 | 14 | 2145 | ||
NM_001163213.1 | NP_001156685.1 | 14 | 2151 | |||
NM_022965.3 | NP_075254.1 | 12 | 1809 | |||
XM_006713868.1 | XP_006713931.1 | 14 | 2170 | |||
XM_006713869.1 | XP_006713932.1 | 14 | 2170 | |||
XM_006713870.1 | XP_006713933.1 | 14 | 2167 | |||
XM_006713871.1 | XP_006713934.1 | 14 | 2164 | |||
XM_006713872.1 | XP_006713935.1 | 14 | 2161 | |||
XM_006713873.1 | XP_006713936.1 | 14 | 2158 | |||
XM_011513420.1 | XP_011511722.1 | 14 | 2164 | |||
XM_011513422.1 | XP_011511724.1 | 14 | 2161 | |||
GenBank mRNA | AB209441.1 | - | 14 | 2153 | ||
AF245114.1 | - | 11 | 1592 | |||
BC121175.2 | - | 7 | 912 | |||
BC128610.1 | - | 7 | 912 | |||
BC153824.1 | - | 7 | 912 | |||
HM244683.1 | - | 14 | 2040 | |||
KJ534834.1 | - | 13 | 1892 | |||
KJ535004.1 | - | 3 | 165 | |||
M58051.1 | - | 13 | 1928 | |||
M59374.1 | - | 1 | 46 | |||
M64347.1 | - | 12 | 1664 |