Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camkv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145621.2 | NP_663596.1 | 8 | 845 | ||
GenBank mRNA | AK147323.1 | - | 8 | 833 | ||
BC017634.1 | - | 8 | 845 | 4527857 | ||
BC022169.1 | - | 6 | 1211 | |||
BC086762.1 | - | 7 | 1043 | |||
BC111033.1 | - | 8 | 840 | 5368225 |