Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ackr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145700.2 | NP_663746.2 | 3 | 582 | ||
GenBank mRNA | AF306532.1 | - | 1 | 131 | ||
AK030643.1 | - | 2 | 148 | |||
AK042430.1 | - | 2 | 253 | |||
AY072796.1 | - | 2 | 833 | |||
AY072938.1 | - | 3 | 874 | |||
BC140999.1 | - | 3 | 582 | 9055611 |