Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145853.2 | NP_665852.1 | 5, 6 | 768 | ||
GenBank mRNA | AF217002.1 | - | 5, 6 | 804 | ||
AK077411.1 | - | 5, 6 | 768 | |||
AK137626.1 | - | 3, 4 | 466 | |||
AK148129.1 | - | 5, 6 | 829 | |||
BC058951.1 | - | 5, 6 | 768 | 6821376 |