Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146242.2 | NP_666354.1 | 1 | 963 | ||
GenBank mRNA | AB091100.1 | - | 1 | 933 | ||
AF527781.1 | - | 1 | 949 | |||
AK084466.1 | - | 1 | 948 | |||
AK085779.1 | - | 1 | 951 | |||
AK142167.1 | - | 1 | 950 | |||
BC086911.1 | - | 1 | 963 | 6752714 |