Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem41b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153525.5 | NP_705745.3 | 6 | 770 | ||
GenBank mRNA | AK029327.1 | - | 6 | 771 | ||
AK033997.1 | - | 6 | 800 | |||
AK035266.1 | - | 7 | 615 | |||
AK035365.1 | - | 4 | 348 | |||
AK038953.1 | - | 6 | 730 | |||
AK046140.1 | - | 1 | 17 | |||
AK053588.1 | - | 6 | 769 | |||
AK075726.1 | - | 6 | 770 | |||
AK078357.1 | - | 6 | 771 | |||
AK090100.1 | - | 6 | 772 | |||
AK139961.1 | - | 6 | 771 | |||
AK150353.1 | - | 6 | 826 | |||
AK153048.1 | - | 6 | 826 | |||
AK165226.1 | - | 6 | 771 | |||
AK166427.1 | - | 6 | 771 | |||
AK166455.1 | - | 6 | 771 | |||
AK166845.1 | - | 5 | 586 | |||
AK167021.1 | - | 6 | 671 | |||
AK172876.1 | - | 6 | 789 | |||
BC026515.1 | - | 6 | 794 | 4910818 | ||
BC027103.1 | - | 6 | 771 | 5360942 | ||
BC086759.1 | - | 1223 |