Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glb1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153803.1 | NP_722498.1 | 19, 20 | 1913 | ||
GenBank mRNA | AK033400.1 | - | 15, 16 | 1496 | ||
AK143057.1 | - | 18, 19 | 1844 | |||
BC038479.1 | - | 19, 20 | 1913 | 4503030 |