Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153807.2 | NP_722502.1 | 2 | 218 | ||
GenBank mRNA | AK034209.1 | - | 2 | 243 | ||
AK153582.1 | - | 2 | 250 | |||
AK169987.1 | - | 2 | 249 | |||
BC018371.1 | - | 2 | 218 | 4210220 | ||
BC063269.1 | - | 2 | 303 | 4159843 |