Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170756.2 | NP_739562.1 | 2 | 392 | ||
GenBank mRNA | AK051741.1 | - | 2 | 338 | ||
AK122364.1 | - | 1 | 456 | |||
AK144826.1 | - | 2 | 364 | |||
AK156188.1 | - | 2 | 393 | |||
AK162743.1 | - | 2 | 392 | |||
AK170356.1 | - | 2 | 393 | |||
AK171240.1 | - | 2 | 366 | |||
AK171664.1 | - | 2 | 370 | |||
AK171852.1 | - | 2 | 382 | |||
AK201572.1 | - | 1 | 121 | |||
AK210472.1 | - | 1 | 120 | |||
BC034597.1 | - | 2 | 381 | 5363513 |