Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172125.2 | NP_742123.2 | 1 | 1921 | ||
GenBank mRNA | AK016558.1 | - | 1 | 2376 | ||
AK161264.1 | - | 1 | 1682 | |||
BC140979.1 | - | 1 | 2013 | 9055591 |