Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekha7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040271.1 | - | 8 | 1087 | ||
AK048843.1 | - | 8 | 1253 | |||
AK146866.1 | - | 1278 | ||||
AK160810.1 | - | 8 | 1167 | |||
BC060678.1 | - | 7 | 1008 | |||
BC084587.1 | - | 8 | 1182 | 3602481 | ||
EU380771.1 | - | 1304 | ||||
EU380772.1 | - | 1362 |