Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172759.3 | NP_766347.1 | 12 | 2091 | ||
GenBank mRNA | AK033563.1 | - | 12 | 2054 | ||
AK077248.1 | - | 12 | 2082 |