Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfrkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172766.3 | NP_766354.2 | 15 | 1670 | ||
GenBank mRNA | AK041206.1 | - | 15 | 1908 | ||
AK049804.1 | - | 1 | 45 | |||
BC033595.1 | - | 15 | 1660 | 4947357 | ||
BC059861.1 | - | 15 | 1717 | |||
BC059900.1 | - | 6 | 1317 | |||
BC060076.1 | - | 15 | 1899 |