Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: L3mbtl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172787.2 | NP_766375.1 | 19 | 1995 | ||
GenBank mRNA | AK030412.1 | - | 6 | 593 | ||
AK045667.1 | - | 19 | 1996 | |||
AK049597.1 | - | 19 | 2030 | |||
AK129443.1 | - | 15 | 1540 | |||
BC082309.1 | - | 18 | 1865 | 30356672 | ||
BC085192.1 | - | 18 | 2100 | 30694487 |