Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btnl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172793.2 | NP_766381.1 | 4 | 680 | ||
GenBank mRNA | AK080918.1 | - | 4 | 664 | ||
AK084528.1 | - | 4 | 681 | |||
AY177687.1 | - | 4 | 647 | |||
BC025540.1 | - | 4 | 658 | |||
BC052556.1 | - | 3 | 593 | |||
BC151155.1 | - | 4 | 637 | 9088043 |