Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slfn9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172796.2 | NP_766384.2 | 5 | 3537 | ||
GenBank mRNA | AK036579.1 | - | 5 | 3538 | ||
AY217035.1 | - | 4 | 3005 | |||
AY261800.1 | - | 5 | 3445 |