Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Malt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172833.2 | NP_766421.1 | 9 | 1239 | ||
GenBank mRNA | AK041919.1 | - | 9 | 1244 | ||
AK085071.1 | - | 9 | 1239 | |||
AK164626.1 | - | 10 | 1354 | |||
BC110487.1 | - | 10 | 1246 | 40048907 | ||
BC110488.1 | - | 9 | 1213 | 40048908 |